Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172775.2 | NP_766363.2 | 5 | 1627 | ||
GenBank mRNA | AB072381.1 | - | 5 | 1471 | ||
BC150701.1 | - | 4 | 1419 | 9087612 |