Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm15455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161816.1 | NP_001155288.1 | 1 | 2548 | ||
GenBank mRNA | BC157984.1 | - | 1 | 2472 | ||
BC157993.1 | - | 1 | 2472 |
Gene Symbol: Rbm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167775.1 | NP_001161247.1 | 22, 23 | 3249 | ||
NM_001167776.1 | NP_001161248.1 | 21, 22 | 2734 | |||
NM_145627.2 | NP_663602.1 | 22, 23 | 2965 | |||
GenBank mRNA | AK089105.1 | - | 21, 22 | 2545 | ||
AK122208.1 | - | 19, 20 | 4665 | |||
AK145991.1 | - | 22, 23 | 2965 | |||
AK153736.1 | - | 22, 23 | 3250 | |||
AK167666.1 | - | 21, 22 | 2712 | |||
BC004674.1 | - | 21, 22 | 2650 | 3500403 |