Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172147.1 | NP_001165618.1 | 6 | 932 | ||
NM_001172148.1 | NP_001165619.1 | 5 | 850 | |||
NM_001290630.1 | NP_001277559.1 | 6 | 922 | |||
NM_153586.2 | NP_705814.1 | 5 | 850 | |||
GenBank mRNA | AK040213.1 | - | 5 | 790 | ||
AK080041.1 | - | 3 | 616 | |||
AK084514.1 | - | 5 | 785 | |||
AK084629.1 | - | 5 | 785 | |||
AK141075.1 | - | 6 | 857 | |||
AK163147.1 | - | 4 | 769 | |||
AK164496.1 | - | 6 | 880 | |||
AK166005.1 | - | 5 | 785 | |||
AK166229.1 | - | 798 | ||||
BC037024.1 | - | 5 | 817 | 5368185 |