Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053108.4 | NP_444338.2 | 1 | 95 | ||
GenBank mRNA | AB013137.1 | - | 1 | 61 | ||
AF109314.1 | - | 1 | 71 | |||
AF276917.1 | - | 1 | 20 | |||
AK013722.1 | - | 1 | 77 | |||
AK153086.1 | - | 1 | 75 | |||
BC012642.1 | - | 1 | 72 | 4206114 | ||
BC054418.1 | - | 1 | 57 | |||
BY272278.1 | - | 1 | 114 |