Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183427.4 | NP_906272.1 | 9 | 1249 | ||
GenBank mRNA | AK135134.1 | - | 9 | 1393 | ||
AK139335.1 | - | 9 | 1662 | |||
BC037046.1 | - | 5 | 616 | |||
BC056342.1 | - | 9 | 1249 | 5708257 | ||
BC068987.1 | - | 9 | 1249 | 5708257 |