Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178713.4 | NP_848828.1 | 3 | 377 | ||
GenBank mRNA | AF510322.1 | - | 3 | 386 | ||
AK050298.1 | - | 3 | 378 | |||
AK143752.1 | - | 3 | 378 | |||
AK165447.1 | - | 2 | 1693 | |||
BC038493.1 | - | 3 | 389 | 4223059 |