Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003717.1 | NP_001003717.1 | 16 | 2187 | ||
NM_175489.3 | NP_780698.2 | 17 | 2224 | |||
GenBank mRNA | AK050244.1 | - | 16 | 2189 | ||
AK173174.1 | - | 5 | 385 | |||
BC141383.1 | - | 17 | 1923 | 9055996 | ||
CF745582.1 | - | 4 | 266 |