Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099633.1 | NP_001093103.1 | 4 | 943 | ||
GenBank mRNA | AK030981.1 | - | 4 | 908 | ||
AK082443.1 | - | 4 | 832 |