Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012309.2 | NP_001012309.1 | 3 | 178 | ||
GenBank mRNA | AK041865.1 | - | 3 | 164 | ||
AK046155.1 | - | 3 | 164 | |||
AK078762.1 | - | 3 | 164 | |||
AK078945.1 | - | 3 | 165 | |||
AK161058.1 | - | 3 | 179 | |||
AK162143.1 | - | 3 | 165 | |||
AK162990.1 | - | 3 | 171 | |||
AK168573.1 | - | 3 | 164 | |||
BC016601.1 | - | 3 | 153 | |||
BC089560.1 | - | 3 | 211 | 30300075 |