Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012309.2 | NP_001012309.1 | 6 | 570 | ||
GenBank mRNA | AK046155.1 | - | 6 | 556 | ||
AK078762.1 | - | 6 | 556 | |||
AK078945.1 | - | 6 | 557 | |||
AK168573.1 | - | 6 | 556 | |||
BC016601.1 | - | 6 | 545 | |||
BC089560.1 | - | 6 | 603 | 30300075 |