Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012309.2 | NP_001012309.1 | 7 | 1060 | ||
GenBank mRNA | AK078762.1 | - | 7 | 1046 | ||
AK078945.1 | - | 7 | 1048 | |||
AK168573.1 | - | 7 | 1046 | |||
BC016601.1 | - | 7 | 1035 | |||
BC089560.1 | - | 7 | 1093 | 30300075 |