Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phospho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153104.3 | NP_694744.1 | 3 | 812 | ||
GenBank mRNA | AJ457190.1 | - | 3 | 825 |
Gene Symbol: Abi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163464.1 | NP_001156936.1 | 8 | 2713 | ||
NM_025659.4 | NP_079935.1 | 8 | 3064 | |||
GenBank mRNA | AK033965.1 | - | 8 | 2786 |