Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178932.1 | NP_849263.1 | 1, 2 | 1615 | ||
GenBank mRNA | AF350445.2 | - | 1, 2 | 1615 | ||
BC150843.1 | - | 1, 2 | 1606 | 9087754 | ||
BC150848.1 | - | 1, 2 | 1606 | 9087759 |