Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sppl2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082535.1 | NP_001076004.1 | 1 | 1427 | ||
NM_199019.2 | NP_950184.2 | 1 | 1427 | |||
GenBank mRNA | AK077122.1 | - | 1 | 1428 | ||
BC090973.1 | - | 1 | 1402 | |||
BC100418.1 | - | 1 | 1400 | 6706317 |