Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcns3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168564.1 | NP_001162036.1 | 3 | 1358 | ||
NM_173417.3 | NP_775593.1 | 3 | 1421 | |||
GenBank mRNA | AK046054.1 | - | 3 | 1423 | ||
AK134631.1 | - | 1 | 266 | |||
AK162957.1 | - | 3 | 1360 | |||
BC132164.1 | - | 2 | 1072 | 40130441 | ||
BC132464.1 | - | 2 | 1072 | 40130741 |