Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011797.2 | NP_035927.1 | 6 | 802 | ||
GenBank mRNA | AB005450.1 | - | 6 | 802 | ||
AK012316.1 | - | 3 | 179 | |||
AK034693.1 | - | 1 | 427 | |||
AK157764.1 | - | 6 | 802 | |||
BC046995.1 | - | 6 | 784 | 6491779 |