Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172152.2 | NP_742164.1 | 10 | 798 | ||
GenBank mRNA | AY156046.1 | - | 10 | 798 |