Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dido1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291432.1 | NP_001278361.1 | 3 | 325 | ||
NM_001291433.1 | NP_001278362.1 | 3 | 377 | |||
NM_011805.3 | NP_035935.2 | 3 | 377 | |||
NM_175551.4 | NP_780760.2 | 3 | 377 | |||
NM_177852.4 | NP_808520.2 | 3 | 325 | |||
GenBank mRNA | AJ238332.1 | - | 3 | 279 | ||
AK014422.1 | - | 3 | 378 | |||
AK042474.1 | - | 3 | 346 | |||
AK044919.1 | - | 2 | 2490 | |||
AK129117.1 | - | 3 | 325 | |||
AY425951.1 | - | 3 | 279 | |||
AY425952.1 | - | 3 | 279 | |||
BC029110.1 | - | 4 | 379 | |||
BC060249.1 | - | 3 | 325 | |||
BC096662.1 | - | 3 | 317 | |||
BC138712.1 | - | 2 | 154 | 8861734 | ||
BC138713.1 | - | 2 | 154 | 8861735 | ||
BC145401.1 | - | 2 | 154 | |||
BY713603.1 | - | 3 | 375 |