Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF145285.1 | - | 5 | 573 | ||
AK076117.1 | - | 5 | 623 | |||
AK145697.1 | - | 5 | 622 | |||
AK149427.1 | - | 5 | 601 | |||
AK151732.1 | - | 5 | 657 | |||
AK151965.1 | - | 5 | 590 | |||
AK152792.1 | - | 5 | 590 | |||
AK153221.1 | - | 5 | 613 | |||
AK153893.1 | - | 5 | 561 | |||
AK167186.1 | - | 5 | 625 | |||
AK172256.1 | - | 5 | 590 | |||
AK193944.1 | - | 4 | 373 | |||
AK194613.1 | - | 1 | 66 | |||
BC048176.1 | - | 5 | 610 | 6825491 | ||
BC119806.1 | - | 5 | 491 | 40107463 | ||
BC122883.1 | - | 5 | 491 | 40107462 | ||
CN527272.1 | - | 5 | 697 | |||
U65313.1 | - | 5 | 573 |