Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172807.4 | NP_766395.2 | 9 | 1642 | ||
GenBank mRNA | AK028532.1 | - | 9 | 1622 | ||
BC037196.1 | - | 5 | 777 | |||
BC119282.1 | - | 9 | 1620 | 8734031 | ||
BC119310.1 | - | 9 | 1620 | 8734059 |