Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177814.4 | NP_808482.2 | 9, 10 | 2228 | ||
GenBank mRNA | AK122265.1 | - | 7, 8 | 940 | ||
AK147213.1 | - | 10, 11 | 2267 | |||
AY356530.1 | - | 8, 9 | 1915 | |||
AY356531.1 | - | 8, 9 | 1915 | |||
AY356532.1 | - | 8, 9 | 1915 | |||
AY356533.1 | - | 8, 9 | 1915 | |||
BC056760.1 | - | 9, 10 | 2228 | 6331997 |