Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047537.1 | - | 9 | 1124 | ||
AK089383.1 | - | 9 | 1088 | |||
AK171139.1 | - | 9 | 1099 | |||
AK195203.1 | - | 1 | 37 | |||
AK195247.1 | - | 1 | 37 | |||
AK210877.1 | - | 2 | 37 | |||
BC053416.1 | - | 8 | 948 | 6533681 | ||
U00978.1 | - | 8 | 843 |