Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047537.1 | - | 13 | 1593 | ||
AK089383.1 | - | 13 | 1559 | |||
AK171139.1 | - | 13 | 1568 | |||
AK219243.1 | - | 2 | 79 | |||
AK219554.1 | - | 2 | 79 | |||
BC053416.1 | - | 12 | 1417 | 6533681 | ||
U00978.1 | - | 12 | 1312 |