Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001490.3 | NP_001001490.1 | 4 | 1222 | ||
GenBank mRNA | AK142510.1 | - | 4 | 1223 | ||
AY612852.1 | - | 1 | 699 | |||
BC119196.1 | - | 1 | 752 | 8733945 | ||
BC119200.1 | - | 1 | 752 | 8733949 |
Set Membership: |
G-protein coupled receptor |