Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011836.3 | NP_035966.2 | 11 | 1980 | ||
GenBank mRNA | AF079520.1 | - | 11 | 1970 | ||
AF083372.1 | - | 11 | 1917 | |||
AK132948.1 | - | 8 | 965 | |||
BC096366.1 | - | 11 | 1980 | 2865309 |