Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mab21l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011839.3 | NP_035969.2 | 1 | 1216 | ||
GenBank mRNA | AF126427.1 | - | 1 | 1557 | ||
AF149877.1 | - | 1 | 958 | |||
AK053645.1 | - | 1 | 1217 | |||
AK144373.1 | - | 1 | 1386 | |||
BC046628.1 | - | 1 | 1182 | 6493460 | ||
BC058730.1 | - | 1 | 1322 |