Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033337.4 | NP_001028509.2 | 8 | 832 | ||
GenBank mRNA | AK078848.1 | - | 8 | 793 | ||
AK133636.1 | - | 7 | 540 | |||
AK200042.1 | - | 2 | 84 | |||
BC024550.1 | - | 8 | 782 | |||
BC030849.1 | - | 8 | 795 | |||
BC132621.1 | - | 8 | 817 | 40130898 | ||
BC132623.1 | - | 8 | 817 | 40130900 |