Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201470.1 | NP_001188399.1 | 10 | 1548 | ||
NM_011864.3 | NP_035994.2 | 11 | 1584 | |||
GenBank mRNA | AF052453.1 | - | 11 | 1565 | ||
AF085144.1 | - | 10 | 1480 | |||
AK128962.1 | - | 10 | 1569 | |||
AK137675.1 | - | 10 | 1474 | |||
BC090997.1 | - | 11 | 1584 | 30121331 |