Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172824.3 | NP_766412.2 | 6 | 604 | ||
GenBank mRNA | AK029725.1 | - | 6 | 605 | ||
AK090294.1 | - | 6 | 511 | |||
AK172014.1 | - | 1 | 858 | |||
BB618371.1 | - | 6 | 506 | |||
BC031204.1 | - | 6 | 500 | 5066612 | ||
BC095999.1 | - | 6 | 500 | |||
CF742918.1 | - | 4 | 337 |