Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172824.3 | NP_766412.2 | 2, 3 | 269 | ||
GenBank mRNA | AK090294.1 | - | 2, 3 | 176 | ||
BB618371.1 | - | 2, 3 | 171 | |||
BC031204.1 | - | 2, 3 | 165 | 5066612 | ||
BC095999.1 | - | 2, 3 | 165 |