Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290707.1 | NP_001277636.1 | 22, 23 | 2657 | ||
NM_011866.2 | NP_035996.2 | 21, 22 | 2611 | |||
GenBank mRNA | AF110507.1 | - | 21, 22 | 2482 | ||
AK039249.1 | - | 22, 23 | 2657 | |||
AK147690.1 | - | 21, 22 | 2529 | |||
AK162221.1 | - | 21, 22 | 2266 | |||
AK162804.1 | - | 19, 20 | 2152 | |||
AK166310.1 | - | 13, 14 | 1623 | |||
AY360383.1 | - | 21, 22 | 2611 | |||
BC113201.1 | - | 21, 22 | 2477 | 40048238 |