Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291275.1 | NP_001278204.1 | 6 | 743 | ||
NM_001291276.1 | NP_001278205.1 | 4 | 614 | |||
NM_011870.5 | NP_036000.1 | 6 | 758 | |||
GenBank mRNA | AB006463.1 | - | 6 | 539 | ||
AF173010.1 | - | 6 | 537 | |||
AK010345.1 | - | 6 | 555 | |||
AK027915.1 | - | 6 | 570 | |||
AK088450.1 | - | 4 | 429 | |||
AK152805.1 | - | 6 | 590 | |||
AK165472.1 | - | 6 | 543 | |||
BC003714.1 | - | 6 | 544 | 3599986 | ||
BC054385.1 | - | 6 | 563 | 6476137 | ||
BP770454.1 | - | 2 | 294 |