Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011876.3 | NP_036006.1 | 7 | 892 | ||
GenBank mRNA | AK002699.1 | - | 6 | 807 | ||
AK084026.1 | - | 7 | 892 | |||
AK087422.1 | - | 6 | 807 | |||
AK155204.1 | - | 7 | 875 | |||
AK157886.1 | - | 7 | 837 | |||
AK166900.1 | - | 7 | 860 | |||
AK170441.1 | - | 7 | 840 | |||
AY267189.1 | - | 7 | 740 | |||
BC003338.1 | - | 7 | 839 | 3257230 | ||
Y17808.1 | - | 7 | 757 |