Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm4951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033767.3 | NP_001028939.1 | 3 | 1097 | ||
GenBank mRNA | AK149556.1 | - | 2 | 989 | ||
AK149583.1 | - | 3 | 1095 | |||
BC139145.1 | - | 2 | 925 | 8862167 | ||
BC139147.1 | - | 2 | 925 | 8862169 |