Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201353.1 | NP_958741.1 | 10 | 1496 | ||
GenBank mRNA | AK141581.1 | - | 10 | 1521 | ||
BC050103.1 | - | 10 | 1397 | 30024098 | ||
BC057070.1 | - | 10 | 1496 | 5695483 |