Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011889.2 | NP_036019.2 | 9, 10 | 1024 | ||
GenBank mRNA | AF104411.1 | - | 9, 10 | 1047 | ||
AK046722.1 | - | 9, 10 | 1117 | |||
AK141915.1 | - | 9, 10 | 951 | |||
AK164861.1 | - | 9, 10 | 1121 | |||
AK169916.1 | - | 9, 10 | 1145 | |||
BC055738.1 | - | 9, 10 | 953 | 6413697 |