Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peli3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172835.3 | NP_766423.3 | 3 | 393 | ||
GenBank mRNA | AK044418.1 | - | 3 | 340 | ||
AK135575.1 | - | 3 | 342 | |||
BC028931.1 | - | 3 | 351 | 5037308 |