Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011894.2 | NP_036024.2 | 5 | 613 | ||
GenBank mRNA | AB016835.1 | - | 5 | 441 | ||
AK043375.1 | - | 658 | ||||
AK161427.1 | - | 4 | 595 | |||
AK165033.1 | - | 5 | 647 | |||
BC018237.1 | - | 5 | 645 | 3482670 | ||
BC053741.1 | - | 5 | 613 | 6336511 |