Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183046.1 | NP_898867.1 | 6 | 670 | ||
GenBank mRNA | AK045067.1 | - | 6 | 668 | ||
AK049225.1 | - | 6 | 668 | |||
AK135856.1 | - | 6 | 673 | |||
AK143629.1 | - | 6 | 615 | |||
BC151061.1 | - | 6 | 636 | 9087974 | ||
BC172148.1 | - | 6 | 636 | 9054818 |