Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028607.1 | NP_082883.1 | 2 | 153 | ||
GenBank mRNA | AK010745.1 | - | 2 | 150 | ||
AK145313.1 | - | 2 | 154 | |||
BC065806.1 | - | 2 | 146 | 1547643 |