Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153128.2 | NP_694768.1 | 3 | 429 | ||
GenBank mRNA | AK034170.1 | - | 3 | 417 | ||
AK165280.1 | - | 3 | 407 | |||
BC034514.1 | - | 3 | 429 | 3157064 |