Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brinp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207583.2 | NP_997466.2 | 8 | 2068 | ||
GenBank mRNA | AK032297.1 | - | 8 | 2072 | ||
AK045801.1 | - | 8 | 1595 | |||
AK122549.1 | - | 8 | 2134 | |||
BC067038.1 | - | 8 | 2873 | |||
BC076572.1 | - | 8 | 2068 | 30363878 |