Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011907.4 | NP_036037.1 | 2 | 362 | ||
GenBank mRNA | AF151108.1 | - | 2 | 349 | ||
BC104384.1 | - | 1 | 327 | 40050180 | ||
BC104385.2 | - | 1 | 327 | 40050181 | ||
BY714289.1 | - | 2 | 364 |