Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083921.1 | NP_001077390.1 | 8 | 1344 | ||
NM_019705.3 | NP_062679.2 | 9 | 1185 | |||
GenBank mRNA | AF124663.1 | - | 7 | 954 | ||
AK085559.1 | - | 8 | 1333 | |||
AK088591.1 | - | 8 | 1306 | |||
AK146176.1 | - | 9 | 1359 | |||
AK188274.1 | - | 2 | 108 | |||
BC032089.1 | - | 1 | 789 | |||
BC034555.1 | - | 9 | 1181 | 4192310 |