Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083921.1 | NP_001077390.1 | 4 | 742 | ||
NM_019705.3 | NP_062679.2 | 5 | 583 | |||
GenBank mRNA | AF124663.1 | - | 3 | 352 | ||
AK085559.1 | - | 4 | 731 | |||
AK146176.1 | - | 5 | 757 | |||
AK166075.1 | - | 4 | 710 | |||
BC034555.1 | - | 5 | 579 | 4192310 |