Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183022.3 | NP_898843.1 | 2, 3 | 1052 | ||
GenBank mRNA | BC046481.1 | - | 2, 3 | 685 | ||
BC055772.1 | - | 2, 3 | 863 | 6808924 |