Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001309.2 | NP_001001309.1 | 29 | 3149 | ||
GenBank mRNA | AF041409.1 | - | 29 | 2880 | ||
AK031326.1 | - | 14 | 1455 | |||
BC037488.1 | - | 4 | 293 | |||
BC137793.1 | - | 30 | 3134 |