Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005420.1 | NP_001005420.1 | 3 | 951 | ||
GenBank mRNA | AK147361.1 | - | 1 | 3547 | ||
AK172993.1 | - | 1 | 709 | |||
BC070440.1 | - | 3 | 951 | 30362439 |