Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp385b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113399.1 | NP_001106870.1 | 4 | 526 | ||
NM_001113400.1 | NP_001106871.1 | 4 | 548 | |||
NM_178723.6 | NP_848838.2 | 6 | 1058 | |||
GenBank mRNA | AK046798.1 | - | 6 | 1059 | ||
AK050293.1 | - | 4 | 549 | |||
AK149352.1 | - | 6 | 1039 | |||
BC132352.1 | - | 4 | 637 | 40130629 | ||
BC138239.1 | - | 4 | 637 | 8861258 |