Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290986.1 | NP_001277915.1 | 2 | 283 | ||
NM_001290987.1 | NP_001277916.1 | 2 | 298 | |||
NM_030234.2 | NP_084510.2 | 2 | 298 | |||
GenBank mRNA | AK020004.1 | - | 2 | 251 | ||
AK031592.1 | - | 2 | 224 | |||
AK051037.1 | - | 2 | 267 | |||
AK162181.1 | - | 2 | 249 | |||
BC117821.1 | - | 2 | 237 | |||
BC117822.1 | - | 2 | 237 | 40092839 | ||
BY720216.1 | - | 2 | 298 |